Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22811-U

Porter Model 9000 Backpressure Regulator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

General description

Specifications
  • Flow Capacity: 0-1000 cc/min
  • Regulation Range: 0-100 psig (0-7.0 kg/cm2)
  • Construction: aluminum body & bonnet, Fairprene 5029A diaphragm, Viton valve seat
Requires a low flow controller (Cat. No. 22834) with a 0-535 cc/min flow element (Cat. No. 22839).

Application

Backpressure regulator may be used to equalize pressure in sample and reference cells, in an experimental study conducted to measure turbulent fluxes of carbon, temperature, and water vapor, in an upland forest, using eddy covariance method.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon exchange and venting anomalies in an upland deciduous forest in northern Wisconsin, USA.
Cook, Bruce D., et al.
Agricultural and Forest Meteorology, 126, 271-295 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service