Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22796

Supelco

Replacement Filter Element

Synonym(s):

FILTER ELEMENT 0.1UM WITH O-RING FOR NITROGEN GENERATOR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

description

O-ring

compatibility

for use with Air Products Chromatographic Nitrogen Generator

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service