Key Documents
22788-U
Supelco
Fritless Purge Sampler Glassware
size 25 mL, Tekmar® 14-2333-024
About This Item
UNSPSC Code:
41115716
NACRES:
NB.21
feature
fritted: no
manufacturer/tradename
Tekmar® 14-2333-024
size
25 mL
application(s)
industrial hygiene
compatibility
for use with Tekmar® 2000/3000
Application
For use with Tekmar 2000/3000 & Velocity XPT Concentrators
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)