Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22752-U

Recorder Pen for Kipp & Zonen Recorders

black ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
NACRES:
SB.54

material

black ink

packaging

pk of 6

nib L

1/4 in. (7 mm)

General description

For use with recorder models BD7, BD8, BD9, BD10, BD11, BD12, BD14, BD14D, BD30, BD31, BD40, and BD41.

Application

Recorder pen may be used in high-precision isotonic angular position transducer and piezoresistive type pressure transducer to study the tissues from airway strips and perfused airway tubes.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Hulsmann et al.
American journal of respiratory and critical care medicine, 149(2 Pt 1), 519-525 (1994-02-01)
The epithelium probably modulates airway smooth muscle responsiveness by producing relaxing factors, by inactivating agonists, or by acting as a physical barrier. In isolated airway strips, however, only a limited modulatory role of the epithelium has been found, and this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service