Key Documents
22716
Swagelok® Reducing Union
Swagelok® 810-6-4, brass, 1/4 in. Swagelok
Synonym(s):
Swagelok® B810-6-4,BRASS REDUC.UNION 1/2"TO1/4" PK/2
About This Item
UNSPSC Code:
41115717
material
brass
packaging
pkg of 2 ea
manufacturer/tradename
Swagelok® 810-6-4
fitting
1/2 in. Swagelok
1/4 in. Swagelok
General description
Nuts and ferrules included.
Reducing unions of Swagelok® type are mostly used as metal column terminators. All original reducing unions have a bore in the centre of about 6mm in length and an internal diameter of 1.2mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
J.F.K. Huber
Instrumentation for High Performance Liquid Chromatography, 84-88 (2011)