Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22710-U

Swagelok® Miniature Quick Connect Stem Assembly

Swagelok® QM2-S-200, brass, 1/8 in. Swagelok fitting

Synonym(s):

Swagelok® QM2-S-200,BRASS MINI QK CONN 1/8" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

material

brass

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® QM2-S-200

fitting

1/8 in. Swagelok fitting

compatibility

for use with miniature Quick Connects body assembly (22712-U)

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service