Key Documents
22705-U
Swagelok® Connector to Female NPT
Swagelok® 400-7-2, brass, 1/4 in. Swagelok, 1/8 in. female NPT
Synonym(s):
Swagelok® 400-7-2,BRASS CONNECTOR 1/4TX1/8P PK2
About This Item
UNSPSC Code:
41112500
NACRES:
SB.54
material
brass
packaging
pkg of 2 ea
manufacturer/tradename
Swagelok® 400-7-2
fitting
1/4 in. Swagelok
1/8 in. female NPT
General description
Nuts and ferrules included.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)