Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22702

Swagelok® Reducer

new

Swagelok® 100-R-4, stainless steel, 1/16 in. Swagelok, for 1/4 in. tube

Synonym(s):

Swagelok® 100-R-4,SS REDUCER 1/16X1/4 EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 100-R-4

fitting

for 1/4 in. tube
1/16 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Stainless steel Swagelok® reducer maybe used, for ending the top and bottom of continuous membrane fermentor separator (CMFS), used for ethanol fermentation and for increasing the performance of a bioreactor.[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Continuous membrane fermentor separator for ethanol fermentation.
Cho, Chung-Won, and Sun-Tak Hwang.
Journal of Membrane Science, 57, 21-42 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service