Key Documents
22701
Swagelok® Reducer
Swagelok® 100-R-4, brass, 1/16 in. Swagelok, for 1/4 in. tube
About This Item
UNSPSC Code:
41115717
material
brass
packaging
pkg of 2 ea
manufacturer/tradename
Swagelok® 100-R-4
fitting
for 1/4 in. tube
1/16 in. Swagelok
General description
Nuts and ferrules included.
Application
Stainless steel Swagelok® reducer maybe used, for ending the top and bottom of continuous membrane fermentor separator (CMFS), used for ethanol fermentation and for increasing the performance of a bioreactor. It may also be used in automated sample purging assembly for determination of water pollutants using dynamic headspace gas chromatographic technique. It may also be used in combination injection port liner-splitters for gas chromatographic analysis of trialkyllead chlorides.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Headspace chromatographic determination of water pollutants.
Otson, Rein, and David T. Williams.
Analytical Chemistry, 54, 942-946 (1982)
High-resolution gas chromatography of trialkyllead chlorides with an inert solvent venting interface for microwave excited helium plasma detection.
Estes, Scott A., Peter C. Uden, and Ramon M. Barnes.
Analytical Chemistry, 53, 1336-1340 (1981)
Continuous membrane fermentor separator for ethanol fermentation.
Cho, Chung-Won, and Sun-Tak Hwang.
Journal of Membrane Science, 57, 21-42 (1991)