Key Documents
22661
Torque Wrench
About This Item
UNSPSC Code:
41100000
material
stainless steel
General description
Take the guesswork out of tightening a septum nut. This helps to prevent leaking septa, excess bleed, and difficult septum penetration.
The handle of the Supelco septum nut torque wrench slips when the preset torque (8 in.-lbs.) is reached. Deepwell socket (9/16 in.) fits over the Supelco septum nut even with a needle guide attached.
Choose from one of the most recent versions:
Certificates of Analysis (COA)