Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22639-U

Supelco

Glass Fritted TD Tube

glass TD tube, fritted, O.D. × L 8 mm × 11 mm, unconditioned, pkg of 1 ea, matrix base material (glass beads)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

glass TD tube

feature

fritted
unconditioned

packaging

pkg of 1 ea

technique(s)

active air sampling: suitable

O.D. × L

8 mm × 11 mm

matrix

base material (glass beads)

application(s)

air monitoring
industrial hygiene

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B Inhalation

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service