22609-U
Merlin Microseal™ System (fits Varian®)
For CP-1177 injector; 1 nut, 1 inlet adapter, 1 o-ring, and 1 General Purpose (3-100 psi) septum
About This Item
General description
Application
Other Notes
Note: Do not use with beveled tips.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)