Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22548

Supelco

Rotameter Tripod Assembly

for use with Modular rotameters

Synonym(s):

TRIPOD FOR FLOWMETER KITS, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

compatibility

for use with Modular rotameters

Looking for similar products? Visit Product Comparison Guide

General description

Use a rotameter tripod assembly to mount a rotameter anywhere.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarka Salajkova et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-15)
Nosocomial infections, which greatly increase morbidity among hospitalized patients, together with growing antibiotic resistance still encourage many researchers to search for novel antimicrobial compounds. Picolinium salts with different lengths of alkyl chains (C12, C14, C16) were prepared by Menshutkin-like reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service