Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2253505

Supelco

Supelcarb Split-Vent Trap

Replacement trap, pkg of 5 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

packaging

pkg of 5 ea

Looking for similar products? Visit Product Comparison Guide

General description

Split vent may be suitable for gaseous CO2 to flow through during On-Line Supercritical fluid extraction-gas chromatography.[1]
This is a very simple strategy to prevent sample vapors from entering the workplace environment! Our Supelcarb Split Vent Trap traps a broad range of organic compounds, and works with typical split vent flow rates of 10-100 mL/min. The Supelcarb specialty carbon adsorbent is engineered to provide twice the trapping capacity of activated charcoal. Additionally, a narrow particle size distribution and spherical shape allows tight packing and less gas channeling than the irregular shape of activated charcoal particles. We recommend replacing the trap every two weeks. This is based on data that shows that breakthrough of the Supelcarb Split Vent Trap occurs after approximately 2 weeks at 65 mL/min, a much longer time than with other traps that use activated charcoal adsorbent.

Legal Information

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supercritical fluid extraction-capillary gas chromatography: On-line coupling with a programmed temperature vaporizer.
Houben, Robert J., et al.
Journal of High Resolution Chromatography, 13, 669-673 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service