Key Documents
22496
Supelco
Supeltex M-1 Packed Column Ferrule, 1/8 in. Column O.D.
1/8 in. O.D. Column, pkg of 10 ea
About This Item
UNSPSC Code:
41121815
packaging
pkg of 10 ea
I.D.
1/8 in.
compatibility
1/8 in. O.D. Column
General description
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.
Material Composition: Ceramic-filled PTFE
Maximum Temperature:
250 °C
Ferrules are suitable for the attachment of the column and other glass components to the system. Flexible graphite ferrules withstand high temperatures and are capable of compression.[1]
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Walter Jennings
Gas Chromatography with Glass Capillary Columns (2012)