Key Documents
22469
Supelco
Fused Silica Ferrule Kit
About This Item
UNSPSC Code:
41121815
O.D.
0.40-0.51 mm
compatibility
for use with Fused Silica Capillary Columns
General description
Kit for Fused Silica Capillary Columns (0.40 mm to 0.51 mm OD): 4 drilled Supeltex M-2A, 4 indented blank Supeltex M-2A, 4 Supeltex M-4 ferrules, plus pin vise drill kit (Cat. No. 23820-U) and instructions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)