Key Documents
22468
Supelco
Wide Bore Fused Silica Capillary Ferrule Kit
About This Item
UNSPSC Code:
41121815
O.D.
0.8-1.19 mm
compatibility
for use with Glass Capillary Columns
General description
Kit for Wide Bore Fused Silica Capillary Columns (0.8 mm to 1.19 mm OD): 4 Supeltex M-1, 4 Supeltex M-2A, 4 Supeltex M-4 ferrules, plus pin vise drill kit (Cat. No. 23820-U) and instructions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)