Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22440-U

Sigma-Aldrich

Jeweler′s Screwdriver Set

Five interchangeable blades and pouch included

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

General description

Jeweler′s Screwdrivers - Five interchangeable blades connect securely to chuck-type knurled handle with swivel top. Three flat blades: 0.055, 0.070 in., 0.100 in.; two Phillips blades: No. 0, No. 1.
Vinyl storage pouch.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service