Key Documents
22404
Supelco
Packed GC FID Cleaning Kit
for use with Varian instruments
About This Item
UNSPSC Code:
12000000
compatibility
for use with Varian instruments
General description
This collection of wire brushes is specially tailored to clean FIDs (and injection ports) that accept 1/4 in. columns. Brass brushes prevent scratching and marring of expensive FID components and save downtime by allowing the detector to be cleaned while hot. Each kit includes two detector brushes, one injection port tube brush, a brass toothbrush (for cleaning jets and other odd surfaces), and a piece of fine emery cloth to clean electrical contacts.
Choose from one of the most recent versions:
Certificates of Analysis (COA)