22355-U
Chart Paper for Recorder
black, chart type, blank
About This Item
Recommended Products
material
black
feature
chart type blank
manufacturer/tradename
Agilent/HP 9270-1134 (OEM)
roll W × L
107 mm × 400 ft
compatibility
for use with Agilent/HP 85B, 3390, 3390A, and 3392A recorders
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service