Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22337

Supelco

Chart Paper for Recorder

blue, chart type, plain, pkg of 2 rolls

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
14111500

material

blue

feature

chart type plain

packaging

pkg of 2 rolls

manufacturer/tradename

Agilent/HP 5080-8800 (OEM)

paper roll W × L

107 mm × 400 ft

compatibility

for use with Agilent/HP 85B, 3390, 3390A, and 3392A recorders

Looking for similar products? Visit Product Comparison Guide

Application

Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.[1]

Other Notes

Equivalent to Manufacturer′s Chart No. 5080-8800.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Porta et al.
Polymers, 12(10) (2020-10-02)
Reduced periodontal support, deriving from chronic inflammatory conditions, such as periodontitis, is one of the main causes of tooth loss. The use of dental implants for the replacement of missing teeth has attracted growing interest as a standard procedure in
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service