Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22299

Precision Sampling syringe needles

new

volume 100-10000 μL, for use with VICI Series A-2, C, D, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.0 in. × 0.029 in. × 0.012 in.

Synonym(s):

REPLACEMENT TAPER NEEDLEA-2, C, D PK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

bevel tip stainless steel needle (point style 2)

description

A-2, C, D

feature

point style beveled point

packaging

pkg of 3 ea

manufacturer/tradename

VICI 943051

needle L × O.D. × I.D.

2.0 in. × 0.029 in. × 0.012 in.

volume

100-10000 μL

compatibility

for use with VICI Series A-2, C, D

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service