Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22124-U

On/Off Toggle Valve

stainless steel, 1/8 in. angle arms

Synonym(s):

Swagelok® 0GS2-A,SS TOGGLE VALVE 1/8X1/8A EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

material

stainless steel

fitting

1/8 in. angle arms

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.
Toggle valve is one of the most simple and widely used industry valve. It′s on and off settings determines whether or not fluid flows past the valve. There are many styles of toggle valve like ball valve, gate valve, plug valve and knife valve.[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Process Plant Equipment: Operation, Control, and Reliability.
Holloway MD
Technology & Engineering, 401-402 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service