Key Documents
22122
Vernier Handle for Fine Metering Valve
About This Item
UNSPSC Code:
41115717
eCl@ss:
32035302
General description
This optional vernier handle attaches to the stem of a fine metering valve, and is more comfortable to turn than the stem itself. The thicker body also allows for more precise control than possible by turning the stem. Additionally, a scale on the handle can be used for visual detection of movement. Order choice of fine metering valve separately.
Application
Fine metering valve with Vernier handle may be used to regulate gas flow to the ion source from the sample bulb, during chemical ionization mass spectrometry analysis of borazine in methane, ethane, n-butane and ammonia.[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Chemical ionization mass spectrometry of borazine.
Francis RP and Solomon JJ.
Journal of the American Chemical Society, 93 (1), 56-61 (1971)