22115
Fine Metering Valve
stainless steel, 1/8 in. angle arms
Synonym(s):
Swagelok® SS2-A,SS METERING VALVE 1/8A EA
About This Item
Recommended Products
material
stainless steel
description
fine adjustment
fitting
1/8 in. angle arms
Looking for similar products? Visit Product Comparison Guide
General description
application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service