Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22088

Swagelok® Bored-Through Union

Swagelok® 200-6BT, stainless steel, 1/8 in.

Synonym(s):

Swagelok® 200-6BT,SS B-THRU UNION 1/8" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 200-6BT

fitting

1/8 in.

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Swagelok® Bored-through union was suitable for connecting paint can and ground-glass joint for carrying out the continuous steam micro-distillation through a segment of PTFE tube.[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Looking into some basic properties of accelerants in fire residues for higher performance in arson analysis.
Yun-Seng G
Journal of Forensic Sciences, 2 (1), 19-26 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service