Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22060-U

Flexible Stainless Steel Hose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

General description

This infinitely flexible, 30 in. (76 cm) × 1/4 in. 316 grade stainless steel hose solves many routing problems. It enables the direction of a gas line to change as needed without the risk of a leak caused from kinked tubing. Tested to 3000 psig (211 kg/cm2) with helium. End are both 1/8 in. male Swagelok fittings. Nuts and ferrules included.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service