Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22034

Swagelok® Caps

Swagelok® 400-C, stainless steel, 1/4 in. male

Synonym(s):

Swagelok® 400C,SS CAPS 1/4" PK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

material

stainless steel

packaging

pkg of 2 ea

manufacturer/tradename

Swagelok® 400-C

fitting

1/4 in. male

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Swagelok® caps maybe used to seal the conditioned sampling tube, during gas chromatographic determination of low concentrations of volatile organic compounds (≤C6) in ambient air using mercuric oxide reduction gas detection (RGD).[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Passive sampling and gas chromatographic determination of low concentrations of reactive hydrocarbons in ambient air with reduction gas detector.
Xu-Liang C and Hewitt N.
Journal of Chromatography A, 648 (1), 191-197 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service