Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22026

Swagelok® Nut

Swagelok® 402-1, stainless steel, 1/4 in. Swagelok

Synonym(s):

Swagelok® 402-1,SS NUTS 1/4" PK5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

material

stainless steel

packaging

pkg of 5 ea

manufacturer/tradename

Swagelok® 402-1

fitting

1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1]

Application

Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.[2]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diffusion denuder for the collection of semivolatile organic compounds.
Krieger MS and Hites RA.
Environmental Science & Technology, 26 (8), 1551-1555 (1992)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service