Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22025

Swagelok® Union

Swagelok® 100-6, brass, 1/16 in.

Synonym(s):

Swagelok® 100-6,BRASS UNION 1/16" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

brass

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 100-6

fitting

1/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

Nut and ferrules included.
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.[1]

Application

Brass Swagelok® union maybe used in determination of n-butylated trialkyllead compounds by gas chromatography with microwave plasma emission detection.[2]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of n-butylated trialkyllead compounds by gas chromatography with microwave plasma emission detection.
Scott E, at.al.
Analytical Chemistry, 54 (14), 2402-2405 (1982)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service