Key Documents
22023
Swagelok® Back Ferrule
Swagelok 104-1, brass, 1/16 in. Swagelok
Synonym(s):
Swagelok® 104-1,BRASS BK FERR 1/16" PK10
About This Item
UNSPSC Code:
41115717
material
brass
packaging
pkg of 10 ea
manufacturer/tradename
Swagelok 104-1
fitting
1/16 in. Swagelok
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1] Glass chromatographic column require brass back ferrule, PTFE front ferrule and brass nuts for adequate sealing.[2]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Walker JQ
Chromatographic Systems: Maintenance and Troubleshooting, 343-344 (2012)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)