Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22017-U

Swagelok® Reducer

Swagelok® 100-R-2, brass, 1/16 in. Swagelok, for 1/8 in. tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

material

brass

packaging

pkg of 2 ea

manufacturer/tradename

Swagelok® 100-R-2

fitting

for 1/8 in. tube
1/16 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Brass Swagelok® reducer maybe used for in gas chromatography system for breath hydrogen analysis.[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An inexpensive gas chromatograph for breath hydrogen analysis.
T A Robb et al.
Clinica chimica acta; international journal of clinical chemistry, 134(1-2), 235-241 (1983-10-31)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service