Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22001

Swagelok® Front Ferrule

Swagelok® 403-1, brass, 1/4 in. Swagelok

Synonym(s):

Swagelok® 403-1,BRASS FRT FERR 1/4" PK20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

material

brass

packaging

pkg of 20 ea

manufacturer/tradename

Swagelok® 403-1

fitting

1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1] Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing.[2]

Application

Brass front ferrule maybe used for sealing the tube ends to increase the lifetime and performance of the permeation tubes as standard gas sources.[3]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Performance of permeation tubes as standard gas sources.
Bernard S
Environmental Science & Technology, 5 (11), 1121-1128 (1971)
Grob RL & Barry EF.
Modern Practice of Gas Chromatography, 522-523 (2004)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service