Key Documents
21971
Swagelok® 90 Degree Male Elbow
Swagelok® 200-2-4, brass, 1/8 in. Swagelok, 1/4 in. male NPT
Synonym(s):
Swagelok® 200-2-4,BRASS ELBOW MALE, 1/8TX1/4P PK2
About This Item
UNSPSC Code:
41115717
material
brass
packaging
pkg of 2 ea
manufacturer/tradename
Swagelok® 200-2-4
fitting
1/4 in. male NPT
1/8 in. Swagelok
General description
Degree elbow is suitable to change direction of a pipe. In general, 45-degree or 90-degree elbows are used. Elbows enforce sudden inner diameter changes and rough edges which lead to turbulence and greater pressure drop. They are suitable for tubing systems. The capability to bend tubing delivers smoother transition.[1]
Nuts and ferrules included.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Thwarting fluid-system leaks.
Cox, John C.
Chemical Processing (2002)