21953-U
Valco® Precision Sampling plunger tips
size 5 mL, plunger tip for 5mL syringe, pkg of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
PTFE
feature
plunger tip for 5mL syringe
packaging
pkg of 2 ea
manufacturer/tradename
VICI 013935
size
5 mL
compatibility
for use with Series A, A-2, C, & D Precision Sampling Syringe
Looking for similar products? Visit Product Comparison Guide
General description
For series A, A-2, C, and D Pressure-Lok Gas syringes. For 25 μL to 250 μL syringes, factory repair only.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service