Key Documents
21934-U
SGE Plunger-Protected Syringe, Fixed Needle
volume 10 μL, needle size 26 ga, needle L 50 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle
needle size
26 ga
feature
syringe type Plunger-protection
packaging
pkg of 1 × 6 ea (21934-U)
manufacturer/tradename
SGE 10F, 2030
needle L
50 mm
volume
10 μL
General description
Extended protection tube from top of the flange to help prevent plunger bending during injection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)