Key Documents
21924
SGE Replacement Plunger for 10R-VA8 Autosampler Syringe (Varian®)
volume 10 μL, for 10R-VA8, for use with Varian 10μL autosampler syringe
About This Item
UNSPSC Code:
41122001
material
PTFE plunger tip
needle size
10 μL
description
replacement plunger
packaging
pkg of 1 ea
manufacturer/tradename
SGE 031218
volume
10 μL
compatibility
for use with Varian 10μL autosampler syringe
General description
Spare plunger for Varian 10μL syringe.
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)