21879-U
Empty Glass GC Column, for Shimadzu™ 7A, 9A, 12A, 14A, 14B, 15A, 16A, and 2014 (On-Column)
L × O.D. × I.D. 6.0 ft (1.8 m) × 5.0 mm × 2.6 mm
About This Item
material
glass column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 5.0 mm × 2.6 mm
column type
empty
compatibility
for use with Shimadzu
General description
- X (length of injector arm) = 13.0 in. (330 mm)
- Y (length of detector arm) = 11.0 in. (279 mm)
- S (span, injector to detector) = 1.57 in. (40 mm)
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)