21739
Empty Glass GC Column, for PerkinElmer® 8000 and 9000 (On-Column)
L × O.D. × I.D. 3.0 ft (0.91 m) × 1/4 in. × 2.0 mm
About This Item
material
glass column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
3.0 ft (0.91 m) × 1/4 in. × 2.0 mm
column type
empty
compatibility
for use with PerkinElmer
General description
- X (length of injector arm) = 12.0 in. (305 mm)
- Y (length of detector arm) = 6.81 in. (173 mm)
- S (span, injector to detector) = 6.5 in. (165 mm)
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)