Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21664

Swagelok® Bored-Through Union

Swagelok® 400-6BT, brass, 1/4 in.

Synonym(s):

Swagelok® 400-6BT BRASS B-THRU UNION 1/4" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

brass

description

bored-through

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 400-6BT

fitting

1/4 in.

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Swagelok® Bored-through union was suitable for connecting paint can and ground-glass joint in order to carry out the continuous steam micro-distillation through a segment of PTFE tube.[1] It may also be suitable for swaging ferrule onto the tubing to show chip-to-tube bonding demonstrated with a five-port silicon microreactor.[2]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Looking into some basic properties of accelerants in fire residues for higher performance in arson analysis.
Giang, Yun-Seng, et al.
Journal of Forensic Sciences, 2, 19-26 (2003)
Edward R Murphy et al.
Lab on a chip, 7(10), 1309-1314 (2007-09-27)
Constructing a microsystem compatible with a large variety of chemistries requires a system design that will be robust in the presence of different compounds and at a wide range of conditions. Although microreactors themselves can accommodate a great span of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service