2162/3
Cone & Plate Viscosity Standard
certified reference material, 100.4 mPa.s (25.00°C)
About This Item
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
ASTM D 2162
100.4 mPa.s(25.00 °C)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
hcodes
pcodes
Hazard Classifications
Asp. Tox. 1
supp_hazards
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service