Key Documents
21588
Supelco
Empty Glass GC Column, for Tracor 540, 560, 565, 570, and 585 (Not On-Column)
L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
About This Item
UNSPSC Code:
41121800
material
glass column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
column type
empty
compatibility
for use with Tracor
General description
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.
Silane treated empty glass column. Fittings are not included.
  • X (length of injector arm) = 8.0 in. (203 mm)
  • Y (length of detector arm) = 8.0 in. (203 mm)
  • S (span, injector to detector) = 6.0 in. (152 mm)
This column fits Tracor 540, 560, 565, 570, and 585 (not on-column injection).
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
The determination of active filter aid adsorption sites by temperature-programmed desorption.
Zhu. Z. Y.
Journal of the American Oil Chemists' Society, 71, 189-194 (1994)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)