21194-U
Empty Glass GC Column, for Varian® 3000, 4000, 6000, and Vista (FID, Inj. A to Det. A, On-Column)
L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 4.0 mm, TightSpec
About This Item
material
glass column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.6 ft (2.0 m) × 1/4 in. × 4.0 mm, TightSpec
column type
empty
compatibility
for use with Varian
General description
- X (length of injector arm) = 9.31 in. (236 mm)
- Y (length of detector arm) = 8.0 in. (203 mm)
- S (span, injector to detector) = 5.5 in. (140 mm)
Application
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)