Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21135

Supelco

Purge/Trap A

for use with OI Analytical 4460

Synonym(s):

PURGE TRAP A, O.I.ANALYTICAL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

application(s)

environmental

compatibility

for use with OI Analytical 4460

Looking for similar products? Visit Product Comparison Guide

General description

Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials, so that a broad range of polar and nonpolar, high and low molecular weight compounds can be trapped in a single tube. Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition. During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds. During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Boey et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(5), 351-360 (2019-01-10)
Modern electron microscopy offers a wide variety of tools to investigate the ultrastructural organization of cells and tissues and to accurately pinpoint intracellular localizations of macromolecules of interest. New volumetric electron microscopy techniques and new instrumentation provide unique opportunities for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service