Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21105

Supelco

GC Stationary Phase

phase OV-17, bottle of 25 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

agency

meets requirements for USP G3

packaging

bottle of 25 g

parameter

0-375 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

OV-17 phase

solubility

chloroform: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients.[1] The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered before choosing a stationary phase. OV-17 is suitable for the separation of drugs, amino acid derivatives, pesticides, phenols and sugar derivatives.[2]
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

OV-17 is suitable as stationary phase for analysis of free phytosterols using gas liquid chromatography.[3]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

536.0 °F

flash_point_c

280 °C

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of free phytosterols by gas chromatography using liquid phase OV-101.
C Grunwald
Journal of chromatography, 44(1), 173-175 (1969-10-14)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service