Key Documents
21043-U
Supelco
Universal Elution Rack for LpDNPH Cartridges
pk of 1
About This Item
UNSPSC Code:
12190000
NACRES:
NB.24
packaging
pk of 1
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with LpDNPH Cartridges
General description
Universal Elution Rack for LpDNPH Cartridges is an accessory for use with LpDNPH cartridges, which are air sampling devices designed for sampling carbonyls (e.g., formaldehyde) in ambient, indoor, workplace, and source atmospheres.
Choose from one of the most recent versions:
Certificates of Analysis (COA)