Key Documents
20985
Valco® Precision Sampling syringe, series D-140
volume 2 μL, needle, RN (removable needle), blunt tip stainless steel needle, needle L × O.D. × I.D. 2.5 in. × 0.019 in. × 0.008 in.
Synonym(s):
SYRINGE,PS-D-140,2.0UL
About This Item
UNSPSC Code:
41122001
material
blunt tip stainless steel needle
description
D-140
feature
needle RN (removable needle)
packaging
pkg of 1 ea
manufacturer/tradename
VICI 140012
needle L × O.D. × I.D.
2.5 in. × 0.019 in. × 0.008 in.
volume
2 μL
General description
D-140 – Removable Needle: A plunger-in-needle design is featured in this GC sample injection syringe. A positive rear stop prevents plunger bending. It fills by retracting the plunger, and features easy maintenance and replaceable parts.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)