Key Documents
20954
Valco® Precision Sampling syringe, series CG-130
volume 5 μL, CG-130, needle type, fixed, bevel tip stainless steel needle (point style 2), needle L 2 in.
Synonym(s):
SYRINGE,PS-CG-130-FN,5UL
About This Item
UNSPSC Code:
41122001
material
bevel tip stainless steel needle (point style 2)
feature
needle type fixed
packaging
pkg of 1 ea
manufacturer/tradename
VICI 130021
needle L
2 in.
volume
5 μL
General description
CG-130: This model has a reinforced plunger syringe with an internal guide and special plunger that eliminates bending
problems.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)