Key Documents
20712
Supelco
GR-2 Septa
diam. 5 mm ( 3/16 in.), pkg of 100 ea
About This Item
UNSPSC Code:
41115717
packaging
pkg of 100 ea
diam.
5 mm ( 3/16 in.)
General description
Septum is a very important component of syringe injection system. They are usually fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene or fluoroelastomers. It is suitable to conveniently introducing the sample without causing the system to leak. Improper septa decomposes under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
These low cost gray silicone rubber septa are designed for routine, isothermal use. For use with 50 °C to 200 °C inlet temperature.
Application
GR-2 septa were soaked in either a control or Pieris mixture for an hour proceeded by bioassays with synthetic scent compounds.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Florian P Schiestl et al.
The New phytologist, 203(1), 257-266 (2014-04-02)
Plant defense against herbivores may compromise attraction of mutualists, yet information remains limited about the mechanisms underlying such signaling tradeoffs. Here, we investigated the effects of foliar herbivory by two herbivore species on defense compounds, floral signaling, pollinator and parasitoid
Robert L. Grob, PhD, Eugene F. Barry, PhD
Modern Practice of Gas Chromatography, 472-472 (2004)
R.L. Grob, M.A. Kaiser
Environmental Problem Solving Using Gas and Liquid Chromatography, 166-166 (2000)