20662-U
Thermogreen® LB-2 Septa, solid discs
diam. 14 mm ( 9/16 in.), pkg of 50 ea
About This Item
Recommended Products
packaging
pkg of 50 ea
parameter
at 100 - 350 °C (operating range)
diam.
14 mm ( 9/16 in.)
Looking for similar products? Visit Product Comparison Guide
General description
- Extremely low bleed over a wide range of inlet temperatures (100 °C to 350 °C)
- Already conditioned, ready to use
- Easier needle penetration and high puncture tolerance (ideal for autosamplers)
- Rubber formulation exclusive to Supelco
Application
- To minimize extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS.[3]
- Thermo-resistant septa is used in GC instrument to avoid carrier gas leaks, extraneous peaks and phthalate contamination.[4]
- Thermo-resistant septa along with thick needle protector is used in GC instruments to avoid damage irreversibly caused by microsyringe.[5]
- It is also used in the influent sampling ports in gas chromatographs.[6]
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service